Product Details

SNP ID
rs35596291
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:145098188 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCAGCAGCTGGCTTCGCCAACG[C/G]CGTTGAACAAGGGTCGCAGCTCAAT
Phenotype
MIM: 601213 MIM: 613745
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ABCE1 PubMed Links

Gene Details

Gene
ABCE1
Gene Name
ATP binding cassette subfamily E member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040876.1 48 UTR 5 NP_001035809.1
NM_002940.2 48 UTR 5 NP_002931.2
Gene
ANAPC10
Gene Name
anaphase promoting complex subunit 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256706.1 48 UTR 5 NP_001243635.1
NM_001256707.1 48 UTR 5 NP_001243636.1
NM_001256708.1 48 Intron NP_001243637.1
NM_001256709.1 48 UTR 5 NP_001243638.1
NM_001256710.1 48 UTR 5 NP_001243639.1
NM_001256711.1 48 UTR 5 NP_001243640.1
NM_001256712.1 48 UTR 5 NP_001243641.1
NM_001318367.1 48 Intron NP_001305296.1
NM_014885.4 48 UTR 5 NP_055700.2
XM_011531524.2 48 Intron XP_011529826.1
XM_011531525.2 48 Missense Mutation CCG,GCG P12A XP_011529827.1
XM_011531526.2 48 Missense Mutation CCG,GCG P12A XP_011529828.1
XM_011531527.2 48 Missense Mutation CCG,GCG P12A XP_011529829.1
XM_011531529.2 48 Intron XP_011529831.1
XM_011531530.2 48 Intron XP_011529832.1
XM_011531531.2 48 Missense Mutation CCG,GCG P12A XP_011529833.1
XM_011531532.2 48 UTR 5 XP_011529834.1
XM_017007638.1 48 Missense Mutation CCG,GCG P12A XP_016863127.1
XM_017007639.1 48 Intron XP_016863128.1
XM_017007640.1 48 Intron XP_016863129.1
XM_017007641.1 48 UTR 5 XP_016863130.1
XM_017007642.1 48 Intron XP_016863131.1
XM_017007643.1 48 Missense Mutation CCG,GCG P12A XP_016863132.1
XM_017007644.1 48 UTR 5 XP_016863133.1

View Full Product Details