Product Details

SNP ID
rs28412229
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:83142429 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGAAAATCTAAAGCTGTTTTTCA[A/T]CTTGTTAAGTGTGAGGTGCCTAAAA
Phenotype
MIM: 609825
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
COQ2 PubMed Links

Gene Details

Gene
COQ2
Gene Name
coenzyme Q2, polyprenyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015697.7 Intron NP_056512.5
XM_011531855.2 Intron XP_011530157.1
XM_011531857.2 Intron XP_011530159.1
XM_011531859.2 Intron XP_011530161.1
XM_011531860.2 Intron XP_011530162.1
XM_011531862.2 Intron XP_011530164.1
XM_011531863.2 Intron XP_011530165.1
XM_011531866.2 Intron XP_011530168.1
XM_011531867.2 Intron XP_011530169.1
XM_017008031.1 Intron XP_016863520.1
XM_017008032.1 Intron XP_016863521.1

View Full Product Details