Product Details

SNP ID
rs28427518
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:165875173 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAGCGAAGCGCAGAGGAAGGGAG[C/T]GGACAAAGTCAAATATATCTAGGGT
Phenotype
MIM: 606742
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TLL1 PubMed Links

Gene Details

Gene
TLL1
Gene Name
tolloid like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204760.1 Intron NP_001191689.1
NM_012464.4 Intron NP_036596.3
XM_011532214.1 Intron XP_011530516.1
XM_017008570.1 Intron XP_016864059.1

View Full Product Details