Product Details

SNP ID
rs28438614
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:116576133 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAAAACCTAGTCTTCACACCTCC[A/G]GGGAGGTAGATATTTCTCTGCACTC
Phenotype
MIM: 153420 MIM: 603491
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD58 PubMed Links

Gene Details

Gene
CD58
Gene Name
CD58 molecule
There are no transcripts associated with this gene.

Gene
IGSF3
Gene Name
immunoglobulin superfamily member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007237.2 5454 UTR 3 NP_001007238.1
NM_001542.3 5454 UTR 3 NP_001533.2
XM_005270793.2 5454 UTR 3 XP_005270850.1
XM_005270794.4 5454 UTR 3 XP_005270851.1
XM_006710593.3 5454 UTR 3 XP_006710656.1
XM_011541315.1 5454 UTR 3 XP_011539617.1
XM_011541316.2 5454 UTR 3 XP_011539618.1

View Full Product Details