Product Details
- SNP ID
-
rs1799940
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:109606350 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACGGAAAACTTGGTTTCAATGATG[T/C]CCGTGGTTTTGACTCTGGATCGGAG
- Phenotype
-
MIM: 139340
MIM: 611189
- Polymorphism
- T/C, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GNAT2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1799875] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GNAT2
- Gene Name
- G protein subunit alpha transducin 2
- Gene
- MIR197
- Gene Name
- microRNA 197
There are no transcripts associated with this gene.
View Full Product Details