Product Details

SNP ID
rs5744196
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:223108680 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATTCCATGGGGAGGAAGAAGCA[C/T]GGGCAGATGTGTAAGTGGGAAACCC
Phenotype
MIM: 603031
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TLR5 PubMed Links

Gene Details

Gene
TLR5
Gene Name
toll like receptor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003268.5 4845 Intron NP_003259.2
XM_005273241.4 4845 UTR 3 XP_005273298.2
XM_005273242.4 4845 UTR 3 XP_005273299.2
XM_005273243.4 4845 UTR 3 XP_005273300.2
XM_006711504.3 4845 UTR 3 XP_006711567.1
XM_006711505.3 4845 UTR 3 XP_006711568.1
XM_006711506.3 4845 UTR 3 XP_006711569.1
XM_011509937.2 4845 UTR 3 XP_011508239.1
XM_017002208.1 4845 UTR 3 XP_016857697.1

View Full Product Details