Product Details

SNP ID
rs29000585
Assay Type
Functionally Tested
NCBI dbSNP Submissions
5
Location
Chr.1:8878923 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCCCGACTCCCCAGCGGCCCTG[G/T]AGTGGTGCGGGCGAAACTCTGGCCC
Phenotype
MIM: 172430
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
ENO1 PubMed Links

Gene Details

Gene
ENO1
Gene Name
enolase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201483.1 170 Intron NP_001188412.1
NM_001428.3 170 UTR 5 NP_001419.1
XM_006710433.2 170 Intron XP_006710496.1
Gene
ENO1-AS1
Gene Name
ENO1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details