Product Details

SNP ID
rs28377971
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:65232251 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACTAGGGTGATTTCATCAGCATT[G/T]AGTTAGATCACCAACAAAATGAAGG
Phenotype
MIM: 604221
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
ACTR2 PubMed Links

Gene Details

Gene
ACTR2
Gene Name
ARP2 actin related protein 2 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005386.2 Intron NP_001005386.1
NM_005722.3 Intron NP_005713.1

View Full Product Details