Product Details

SNP ID
rs28590093
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:84462483 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCTGAGTCACGAGTGCATCCTT[A/T]ACCTTGGCAAAATAAATTTTCTAAA
Phenotype
MIM: 603336 MIM: 611224
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DNAH6 PubMed Links

Gene Details

Gene
DNAH6
Gene Name
dynein axonemal heavy chain 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001370.1 Intron NP_001361.1
XM_006711956.2 Intron XP_006712019.1
XM_011532649.2 Intron XP_011530951.1
XM_011532650.2 Intron XP_011530952.1
XM_011532652.1 Intron XP_011530954.1
XM_011532653.1 Intron XP_011530955.1
XM_011532654.1 Intron XP_011530956.1
XM_011532655.2 Intron XP_011530957.1
XM_011532657.1 Intron XP_011530959.1
XM_011532660.1 Intron XP_011530962.1
XM_017003521.1 Intron XP_016859010.1
XM_017003522.1 Intron XP_016859011.1
XM_017003523.1 Intron XP_016859012.1
XM_017003524.1 Intron XP_016859013.1
Gene
SUCLG1
Gene Name
succinate-CoA ligase alpha subunit
There are no transcripts associated with this gene.

View Full Product Details