Product Details

SNP ID
rs34318454
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:10032474 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGTGTGGAGGGGGGGAATGGGG[A/G]TGAAGGGGGGGAGATGGGGGGGTGG
Phenotype
MIM: 613984
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CIDECP PubMed Links

Gene Details

Gene
CIDECP
Gene Name
cell death-inducing DFFA-like effector c pseudogene
There are no transcripts associated with this gene.

Gene
FANCD2
Gene Name
Fanconi anemia complementation group D2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018115.2 Intron NP_001018125.1
NM_001319984.1 Intron NP_001306913.1
NM_033084.4 Intron NP_149075.2

View Full Product Details