Product Details

SNP ID
rs34242752
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:10082249 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCTCCACCGTCTTTTTCCCACAT[C/G]TGATTGCCAAATCCTGTTGATTTTA
Phenotype
MIM: 613984
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FANCD2 PubMed Links

Gene Details

Gene
FANCD2
Gene Name
Fanconi anemia complementation group D2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018115.2 Intron NP_001018125.1
NM_001319984.1 Intron NP_001306913.1
NM_033084.4 Intron NP_149075.2
Gene
FANCD2OS
Gene Name
FANCD2 opposite strand
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164839.1 Intron NP_001158311.1
NM_173472.1 Intron NP_775743.1
XM_011533330.2 Intron XP_011531632.1
XM_011533331.2 Intron XP_011531633.1

View Full Product Details