Product Details

SNP ID
rs34391207
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:23916990 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTGCGGAGCAGGGGGCGGGACAA[C/T]AGCGGCCGCGGCGCCCCACTCGGCA
Phenotype
MIM: 604496 MIM: 604174
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NKIRAS1 PubMed Links

Gene Details

Gene
NKIRAS1
Gene Name
NFKB inhibitor interacting Ras like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020345.3 187 UTR 5 NP_065078.1
XM_005265076.3 187 Intron XP_005265133.1
XM_005265077.4 187 UTR 5 XP_005265134.1
XM_005265078.3 187 UTR 5 XP_005265135.1
Gene
RPL15
Gene Name
ribosomal protein L15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253379.1 187 UTR 5 NP_001240308.1
NM_001253380.1 187 Intron NP_001240309.1
NM_001253382.1 187 Intron NP_001240311.1
NM_001253383.1 187 Intron NP_001240312.1
NM_001253384.1 187 UTR 5 NP_001240313.1
NM_002948.3 187 UTR 5 NP_002939.2

View Full Product Details