Product Details
- SNP ID
-
rs28362640
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:39108376 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCCCTCCCATCTGGTTTCTCTGCT[C/G]TGGCTCTTTTCAGCATCAGTAGTCC
- Phenotype
-
MIM: 606867
MIM: 611430
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GORASP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112193584] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GORASP1
- Gene Name
- golgi reassembly stacking protein 1
- Gene
- TTC21A
- Gene Name
- tetratricopeptide repeat domain 21A
View Full Product Details