Product Details

SNP ID
rs35588362
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:177312341 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATAGCTCCTCTCAGCCATAAAAG[A/T]TTCAGTAGGATCTTTATTGCTTTTT
Phenotype
MIM: 608934
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NEIL3 PubMed Links

Gene Details

Gene
NEIL3
Gene Name
nei like DNA glycosylase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018248.2 Intron NP_060718.2
XM_017008360.1 Intron XP_016863849.1

View Full Product Details