Product Details

SNP ID
rs28627698
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:181064213 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTTTGTGTTTTTAAAATATTCA[C/T]AATGCCATTCATGAATCACTCTACG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00290 PubMed Links

Gene Details

Gene
LINC00290
Gene Name
long intergenic non-protein coding RNA 290
There are no transcripts associated with this gene.

View Full Product Details