Product Details
- SNP ID
-
rs28670063
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:169630222 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATGCTGCGGTTTGGGGTACAGTTG[C/T]ACCCATTACCCAGGAAGTGGGTATA
- Phenotype
-
MIM: 600580
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CLCN3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113109873] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CLCN3
- Gene Name
- chloride voltage-gated channel 3
View Full Product Details