Product Details

SNP ID
rs28629788
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:177523822 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGGCTGGTGGGAGAGGGCCTGG[C/G]GGGGCGGTGAGCCTCCCCAAGGGGT
Phenotype
MIM: 608170 MIM: 615813
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DDX41 PubMed Links

Gene Details

Gene
DDX41
Gene Name
DEAD-box helicase 41
There are no transcripts associated with this gene.

Gene
FAM193B
Gene Name
family with sequence similarity 193 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190946.2 Intron NP_001177875.1
XM_006714875.2 Intron XP_006714938.1
XM_006714876.1 Intron XP_006714939.1
XM_006714878.1 Intron XP_006714941.1
XM_006714879.3 Intron XP_006714942.1
XM_006714880.1 Intron XP_006714943.1
XM_006714884.1 Intron XP_006714947.1
XM_006714885.1 Intron XP_006714948.1
XM_006714886.2 Intron XP_006714949.1
XM_011534572.1 Intron XP_011532874.1
XM_011534573.1 Intron XP_011532875.1
XM_011534574.1 Intron XP_011532876.1
XM_011534579.2 Intron XP_011532881.1
XM_011534580.1 Intron XP_011532882.1
XM_011534581.1 Intron XP_011532883.1
XM_011534582.1 Intron XP_011532884.1
XM_011534583.1 Intron XP_011532885.1

View Full Product Details