Product Details

SNP ID
rs28449232
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:177602233 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGGGGTTTCCTACCAGGCCACC[C/T]CTCAGCTGCGTGTGGAACCTGGGCC
Phenotype
MIM: 604327
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
B4GALT7 PubMed Links

Gene Details

Gene
B4GALT7
Gene Name
beta-1,4-galactosyltransferase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007255.2 1135 Intron NP_009186.1
XM_006714816.3 1135 Intron XP_006714879.1
XM_017008999.1 1135 UTR 5 XP_016864488.1
Gene
TMED9
Gene Name
transmembrane p24 trafficking protein 9
There are no transcripts associated with this gene.

View Full Product Details