Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001135044.1 | 3302 | Intron | NP_001128516.1 | ||
NM_001308244.1 | 3302 | Intron | NP_001295173.1 | ||
NM_002752.4 | 3302 | UTR 3 | NP_002743.3 | ||
NM_139068.2 | 3302 | UTR 3 | NP_620707.1 | ||
NM_139069.2 | 3302 | UTR 3 | NP_620708.1 | ||
NM_139070.2 | 3302 | UTR 3 | NP_620709.1 | ||
XM_005265940.4 | 3302 | UTR 3 | XP_005265997.1 | ||
XM_006714891.2 | 3302 | UTR 3 | XP_006714954.1 | ||
XM_017009638.1 | 3302 | UTR 3 | XP_016865127.1 | ||
XM_017009639.1 | 3302 | UTR 3 | XP_016865128.1 | ||
XM_017009640.1 | 3302 | UTR 3 | XP_016865129.1 | ||
XM_017009641.1 | 3302 | UTR 3 | XP_016865130.1 | ||
XM_017009642.1 | 3302 | Intron | XP_016865131.1 | ||
XM_017009643.1 | 3302 | Intron | XP_016865132.1 |