Product Details

SNP ID
rs28690799
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:24443303 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATTTCTTCTGTCCACCATTAGAG[C/G]AGCCAGTTTTTCAAGTTCTCACTTG
Phenotype
MIM: 607310
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ADAM7 PubMed Links

Gene Details

Gene
ADAM7
Gene Name
ADAM metallopeptidase domain 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003817.3 Intron NP_003808.2
XM_017013943.1 Intron XP_016869432.1
Gene
LOC101929294
Gene Name
uncharacterized LOC101929294
There are no transcripts associated with this gene.

View Full Product Details