Product Details

SNP ID
rs28517329
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:27286569 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAGGCCTGGCAAGGAAATAGGCC[A/G]AAATCACGATTTAAAAAATGTTGTG
Phenotype
MIM: 617007
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM35 PubMed Links

Gene Details

Gene
TRIM35
Gene Name
tripartite motif containing 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304495.1 2575 UTR 3 NP_001291424.1
NM_171982.4 2575 UTR 3 NP_741983.2
XM_005273452.3 2575 UTR 3 XP_005273509.1
XM_011544453.1 2575 UTR 3 XP_011542755.1

View Full Product Details