Product Details

SNP ID
rs28549706
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144054107 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATCCTCTCTGCACGTGGGCACAG[C/T]GGGAGCTCAGCGCAGGCCCAGCTGA
Phenotype
MIM: 606491 MIM: 614243 MIM: 610874
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EXOSC4 PubMed Links

Gene Details

Gene
EXOSC4
Gene Name
exosome component 4
There are no transcripts associated with this gene.

Gene
MIR6846
Gene Name
microRNA 6846
There are no transcripts associated with this gene.

Gene
OPLAH
Gene Name
5-oxoprolinase (ATP-hydrolysing)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017570.4 Intron NP_060040.1
XM_011516960.1 Intron XP_011515262.1
Gene
SMPD5
Gene Name
sphingomyelin phosphodiesterase 5
There are no transcripts associated with this gene.

Gene
SPATC1
Gene Name
spermatogenesis and centriole associated 1
There are no transcripts associated with this gene.

View Full Product Details