Product Details

SNP ID
rs28642716
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:31137057 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGACAAAAGTCAGTTTCATCCTC[A/G]GTGATGCAAATGGTGAGACTGCACA
Phenotype
MIM: 610703
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSPH1 PubMed Links

Gene Details

Gene
HSPH1
Gene Name
heat shock protein family H (Hsp110) member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286503.1 2819 UTR 3 NP_001273432.1
NM_001286504.1 2819 UTR 3 NP_001273433.1
NM_001286505.1 2819 UTR 3 NP_001273434.1
NM_006644.3 2819 UTR 3 NP_006635.2
XM_005266236.1 2819 UTR 3 XP_005266293.1
XM_011534887.2 2819 UTR 3 XP_011533189.1
XM_011534888.1 2819 UTR 3 XP_011533190.1
XM_017020361.1 2819 UTR 3 XP_016875850.1
XM_017020362.1 2819 UTR 3 XP_016875851.1
XM_017020363.1 2819 UTR 3 XP_016875852.1
XM_017020364.1 2819 UTR 3 XP_016875853.1

View Full Product Details