Product Details

SNP ID
rs28365891
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81513276 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCCCTCGGGCTCCACGAGGATC[C/T]CGGGCGCCAGTGCCGGGCTGAGGAA
Phenotype
MIM: 102560 MIM: 607643
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACTG1 PubMed Links

Gene Details

Gene
ACTG1
Gene Name
actin gamma 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199954.1 258 Intron NP_001186883.1
NM_001614.3 258 Intron NP_001605.1
Gene
FSCN2
Gene Name
fascin actin-bundling protein 2, retinal
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077182.2 258 Intron NP_001070650.1
NM_012418.3 258 Intron NP_036550.1
XM_011524587.2 258 Intron XP_011522889.1
XM_011524590.2 258 UTR 5 XP_011522892.1

View Full Product Details