Product Details

SNP ID
rs28538630
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:77253600 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGTTAATTTCAGGGCTGTCATGG[A/G]TAGGTGGAGATTTCAGTTATTTCAA
Phenotype
MIM: 600377
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GALR1 PubMed Links

Gene Details

Gene
GALR1
Gene Name
galanin receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001480.3 Intron NP_001471.2
XM_017025691.1 Intron XP_016881180.1

View Full Product Details