Product Details
- SNP ID
-
rs35737142
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:22040964 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTCAACTGCAGTTACACAGTCAGC[C/G]GTTTAAGAGGGCTGTTCTGGTATAG
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TRAV20
PubMed Links
Gene Details
- Gene
- TRAV20
- Gene Name
- T cell receptor alpha variable 20
There are no transcripts associated with this gene.
View Full Product Details