Product Details

SNP ID
rs28469649
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:64505531 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGTGGAAAAGATCTGTTTCTTGG[C/G]AAGGGAGTGCTTTCGCTTGCAGCGG
Phenotype
MIM: 616578 MIM: 194541
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
LOC102723809 PubMed Links
Additional Information
For this assay, SNP(s) [rs113044684] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
LOC102723809
Gene Name
uncharacterized LOC102723809
There are no transcripts associated with this gene.

Gene
ZBTB1
Gene Name
zinc finger and BTB domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001123329.1 Intron NP_001116801.1
NM_014950.2 Intron NP_055765.2
XM_005267410.1 Intron XP_005267467.1
XM_011536566.2 Intron XP_011534868.1
XM_011536568.2 Intron XP_011534870.1
XM_017021094.1 Intron XP_016876583.1
XM_017021095.1 Intron XP_016876584.1
Gene
ZBTB25
Gene Name
zinc finger and BTB domain containing 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304507.1 Intron NP_001291436.1
NM_001304508.1 Intron NP_001291437.1
NM_006977.3 Intron NP_008908.2
XM_005268051.3 Intron XP_005268108.1
XM_006720250.3 Intron XP_006720313.1
XM_006720251.3 Intron XP_006720314.1
XM_006720252.3 Intron XP_006720315.1
XM_011537139.1 Intron XP_011535441.1
XM_011537141.2 Intron XP_011535443.1
XM_011537143.2 Intron XP_011535445.1
XM_011537146.2 Intron XP_011535448.1
XM_017021630.1 Intron XP_016877119.1
XM_017021631.1 Intron XP_016877120.1
XM_017021632.1 Intron XP_016877121.1
XM_017021633.1 Intron XP_016877122.1
XM_017021634.1 Intron XP_016877123.1
XM_017021635.1 Intron XP_016877124.1
XM_017021636.1 Intron XP_016877125.1
XM_017021637.1 Intron XP_016877126.1
XM_017021638.1 Intron XP_016877127.1

View Full Product Details