Product Details

SNP ID
rs28675017
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:71330773 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTGTCTATCTCGGGCAAGGCCC[A/G]GTGGGCTGTCCCTGACGACCTCAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SIPA1L1 PubMed Links

Gene Details

Gene
SIPA1L1
Gene Name
signal induced proliferation associated 1 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284245.1 Intron NP_001271174.1
NM_001284246.1 Intron NP_001271175.1
NM_001284247.1 Intron NP_001271176.1
NM_015556.2 Intron NP_056371.1
XM_005267516.4 Intron XP_005267573.1
XM_006720111.2 Intron XP_006720174.1
XM_011536630.1 Intron XP_011534932.1
XM_011536631.1 Intron XP_011534933.1
XM_011536632.1 Intron XP_011534934.1
XM_011536635.2 Intron XP_011534937.1
XM_011536638.2 Intron XP_011534940.1
XM_017021159.1 Intron XP_016876648.1
XM_017021160.1 Intron XP_016876649.1
XM_017021161.1 Intron XP_016876650.1
XM_017021162.1 Intron XP_016876651.1
XM_017021163.1 Intron XP_016876652.1
XM_017021164.1 Intron XP_016876653.1
XM_017021165.1 Intron XP_016876654.1
XM_017021166.1 Intron XP_016876655.1
XM_017021167.1 Intron XP_016876656.1
XM_017021168.1 Intron XP_016876657.1
XM_017021169.1 Intron XP_016876658.1
XM_017021170.1 Intron XP_016876659.1
XM_017021171.1 Intron XP_016876660.1
XM_017021172.1 Intron XP_016876661.1
XM_017021173.1 Intron XP_016876662.1
XM_017021174.1 Intron XP_016876663.1
XM_017021175.1 Intron XP_016876664.1
XM_017021176.1 Intron XP_016876665.1
XM_017021177.1 Intron XP_016876666.1
XM_017021178.1 Intron XP_016876667.1
XM_017021179.1 Intron XP_016876668.1
XM_017021180.1 Intron XP_016876669.1
XM_017021181.1 Intron XP_016876670.1
XM_017021182.1 Intron XP_016876671.1
XM_017021183.1 Intron XP_016876672.1
XM_017021184.1 Intron XP_016876673.1
XM_017021185.1 Intron XP_016876674.1
XM_017021186.1 Intron XP_016876675.1
XM_017021187.1 Intron XP_016876676.1
XM_017021188.1 Intron XP_016876677.1
XM_017021189.1 Intron XP_016876678.1
XM_017021190.1 Intron XP_016876679.1
XM_017021191.1 Intron XP_016876680.1
XM_017021192.1 Intron XP_016876681.1
XM_017021193.1 Intron XP_016876682.1
XM_017021194.1 Intron XP_016876683.1
XM_017021195.1 Intron XP_016876684.1
XM_017021196.1 Intron XP_016876685.1
XM_017021197.1 Intron XP_016876686.1
XM_017021198.1 Intron XP_016876687.1
XM_017021199.1 Intron XP_016876688.1
XM_017021200.1 Intron XP_016876689.1

View Full Product Details