Product Details

SNP ID
rs28828113
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:23944098 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTGCGGGATCTGGCCAGCAGCCC[A/G]CAATGCAACAGGGCTCTCTCTTAGT
Phenotype
MIM: 611596 MIM: 616925
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DHRS4 PubMed Links

Gene Details

Gene
DHRS4
Gene Name
dehydrogenase/reductase 4
There are no transcripts associated with this gene.

Gene
DHRS4-AS1
Gene Name
DHRS4 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details