Product Details

SNP ID
rs11552875
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24145877 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAAATCACTGGGTCCAAGACTTG[C/T]AAATGAGGAAACTGAAGACCTGAAA
Phenotype
MIM: 600654 MIM: 602161 MIM: 612487
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EMC9 PubMed Links

Gene Details

Gene
EMC9
Gene Name
ER membrane protein complex subunit 9
There are no transcripts associated with this gene.

Gene
MIR7703
Gene Name
microRNA 7703
There are no transcripts associated with this gene.

Gene
PSME1
Gene Name
proteasome activator subunit 1
There are no transcripts associated with this gene.

Gene
PSME2
Gene Name
proteasome activator subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002818.2 836 Intron NP_002809.2
XM_006720213.1 836 UTR 5 XP_006720276.1
Gene
RNF31
Gene Name
ring finger protein 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001310332.1 836 Intron NP_001297261.1
NM_017999.4 836 Intron NP_060469.4

View Full Product Details