Product Details

SNP ID
rs28757060
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:75014908 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGCTCTTACAGCTAAGCCTTCA[A/G]CAAACCAGTTAACAGCTCCGAGCTT
Phenotype
MIM: 606454 MIM: 604395
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EIF2B2 PubMed Links

Gene Details

Gene
EIF2B2
Gene Name
eukaryotic translation initiation factor 2B subunit beta
There are no transcripts associated with this gene.

Gene
MLH3
Gene Name
mutL homolog 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040108.1 6575 UTR 3 NP_001035197.1
NM_014381.2 6575 UTR 3 NP_055196.2
XM_005267532.4 6575 UTR 3 XP_005267589.1
XM_005267533.4 6575 UTR 3 XP_005267590.1
XM_005267534.2 6575 Intron XP_005267591.1
XM_006720116.3 6575 Intron XP_006720179.1
XM_011536646.2 6575 UTR 3 XP_011534948.1
XM_011536648.2 6575 Intron XP_011534950.1
XM_017021219.1 6575 UTR 3 XP_016876708.1

View Full Product Details