Product Details

SNP ID
rs28628823
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:76964173 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTCCTGCACAAACCATGGTCCCC[A/C]CTGTTCCCTACAGGAGGAAGAGTTG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LINC01629 PubMed Links

Gene Details

Gene
LINC01629
Gene Name
long intergenic non-protein coding RNA 1629
There are no transcripts associated with this gene.

View Full Product Details