Product Details

SNP ID
rs28699349
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:77800293 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACGCCGGGAGTCGGCCTCTGCTCC[G/T]GTTGTGCAGCCGCCCTGGGCGCCGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ADCK1 PubMed Links

Gene Details

Gene
ADCK1
Gene Name
aarF domain containing kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142545.1 Intron NP_001136017.1
NM_020421.3 Intron NP_065154.2
XM_005267885.3 Intron XP_005267942.1
XM_005267886.3 Intron XP_005267943.1
XM_011536985.1 Intron XP_011535287.1
XM_011536987.2 Intron XP_011535289.1
XM_011536988.2 Intron XP_011535290.1
XM_017021474.1 Intron XP_016876963.1
XM_017021475.1 Intron XP_016876964.1
XM_017021476.1 Intron XP_016876965.1
XM_017021477.1 Intron XP_016876966.1

View Full Product Details