Product Details

SNP ID
hCV63317616
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:92939768 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGCAGACTGGGATTGAAATTTT[A/T]TTTTTAAAAGGTAAAGCTGTTTTAT
Phenotype
MIM: 118910 MIM: 601838
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CHGA PubMed Links

Gene Details

Gene
CHGA
Gene Name
chromogranin A
There are no transcripts associated with this gene.

Gene
ITPK1
Gene Name
inositol-tetrakisphosphate 1-kinase
There are no transcripts associated with this gene.

View Full Product Details