Product Details

SNP ID
rs34699437
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:102933160 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTCCTGGGCCTTTGTCCATGAC[C/T]AGGTGGTCAGCAGGACTTCTGCAGC
Phenotype
MIM: 605799 MIM: 614062
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AMN PubMed Links

Gene Details

Gene
AMN
Gene Name
amnion associated transmembrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030943.3 5920 Intron NP_112205.2
Gene
CDC42BPB
Gene Name
CDC42 binding protein kinase beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006035.3 5920 UTR 3 NP_006026.3
XM_005268227.1 5920 UTR 3 XP_005268284.1
XM_005268228.1 5920 UTR 3 XP_005268285.1
XM_005268229.1 5920 UTR 3 XP_005268286.1
XM_005268230.4 5920 Intron XP_005268287.1
XM_011537387.1 5920 Intron XP_011535689.1

View Full Product Details