Product Details

SNP ID
rs34641434
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:102934658 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGCACTAGGAATAGAGGGGAAC[C/T]TCCTCAACAAAATAAAACAGCTAAC
Phenotype
MIM: 605799 MIM: 614062
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AMN PubMed Links

Gene Details

Gene
AMN
Gene Name
amnion associated transmembrane protein
There are no transcripts associated with this gene.

Gene
CDC42BPB
Gene Name
CDC42 binding protein kinase beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006035.3 Intron NP_006026.3
XM_005268227.1 Intron XP_005268284.1
XM_005268228.1 Intron XP_005268285.1
XM_005268229.1 Intron XP_005268286.1
XM_005268230.4 Intron XP_005268287.1
XM_011537387.1 Intron XP_011535689.1

View Full Product Details