Product Details
- SNP ID
-
rs11556803
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:49837914 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACAATAGAAGAATGCAAAAACAAA[A/C]AAACAAAAAAACAGAAAATAGATTC
- Phenotype
-
MIM: 615104
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
BCDIN3D
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112369913] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BCDIN3D
- Gene Name
- BCDIN3 domain containing RNA methyltransferase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_181708.2 |
1378 |
UTR 3 |
|
|
NP_859059.1 |
- Gene
- BCDIN3D-AS1
- Gene Name
- BCDIN3D antisense RNA 1
There are no transcripts associated with this gene.
- Gene
- NCKAP5L
- Gene Name
- NCK associated protein 5 like
There are no transcripts associated with this gene.
View Full Product Details