Product Details

SNP ID
rs35185648
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53102603 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACACTGATCTTTGGACACAAAGG[G/T]AGGGGAAGGATTAATAGTTAATCCC
Phenotype
MIM: 146735 MIM: 601311
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IGFBP6 PubMed Links

Gene Details

Gene
IGFBP6
Gene Name
insulin like growth factor binding protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002178.2 167 Intron NP_002169.1
XM_017019264.1 167 Intron XP_016874753.1
Gene
SOAT2
Gene Name
sterol O-acyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003578.3 167 Intron NP_003569.1
XM_011538849.2 167 UTR 5 XP_011537151.1
XM_011538850.2 167 UTR 5 XP_011537152.1
XM_011538851.2 167 UTR 5 XP_011537153.1

View Full Product Details