Product Details

SNP ID
rs34745448
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53973938 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCGCCCCCAGTAGGTAGCAGCGG[C/T]CGGGGAACGGGCGGGCAGCGAGGGA
Phenotype
MIM: 611400 MIM: 605559
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HOTAIR PubMed Links

Gene Details

Gene
HOTAIR
Gene Name
HOX transcript antisense RNA
There are no transcripts associated with this gene.

Gene
HOXC11
Gene Name
homeobox C11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014212.3 Intron NP_055027.1

View Full Product Details