Product Details

SNP ID
rs3210158
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56160263 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGGTGAGAAGATGACAGAGGAA[A/G]AAGTAGAGATGCTGGTGGCAGGGCA
Phenotype
MIM: 609931 MIM: 609930 MIM: 601734
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LOC107987175 PubMed Links

Gene Details

Gene
LOC107987175
Gene Name
uncharacterized LOC107987175
There are no transcripts associated with this gene.

Gene
MYL6
Gene Name
myosin light chain 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021019.4 511 Missense Mutation AAA,GAA K124E NP_066299.2
NM_079423.3 511 Missense Mutation AAA,GAA K124E NP_524147.2
Gene
MYL6B
Gene Name
myosin light chain 6B
There are no transcripts associated with this gene.

Gene
SMARCC2
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin subfamily c member 2
There are no transcripts associated with this gene.

View Full Product Details