Product Details

SNP ID
rs34405742
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57129506 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGCCAGGCCAGGGGTACCCTGC[C/T]GGGCAGAGGGCAGGCAGGCCCGGGA
Phenotype
MIM: 107770
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRP1 PubMed Links

Gene Details

Gene
LRP1
Gene Name
LDL receptor related protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002332.2 Intron NP_002323.2
XM_017019303.1 Intron XP_016874792.1

View Full Product Details