Product Details

SNP ID
rs28382833
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57521558 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAAAAGGGGCCGAGGTACTGGAAT[C/T]CCTTGGTAGTCATTTTCCGCTTCTC
Phenotype
MIM: 607376 MIM: 126337 MIM: 156560 MIM: 614489
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCTN2 PubMed Links

Gene Details

Gene
DCTN2
Gene Name
dynactin subunit 2
There are no transcripts associated with this gene.

Gene
DDIT3
Gene Name
DNA damage inducible transcript 3
There are no transcripts associated with this gene.

Gene
MARS
Gene Name
methionyl-tRNA synthetase
There are no transcripts associated with this gene.

Gene
MBD6
Gene Name
methyl-CpG binding domain protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052897.3 613 Intron NP_443129.3
XM_005268617.1 613 Intron XP_005268674.1
XM_005268618.1 613 Intron XP_005268675.1
XM_005268620.2 613 Intron XP_005268677.1
XM_006719217.1 613 Intron XP_006719280.1
XM_006719218.1 613 Intron XP_006719281.1
XM_011537851.2 613 UTR 5 XP_011536153.1
Gene
MIR616
Gene Name
microRNA 616
There are no transcripts associated with this gene.

Gene
MIR6758
Gene Name
microRNA 6758
There are no transcripts associated with this gene.

View Full Product Details