Product Details
- SNP ID
-
rs199827793
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:10453657 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTAGTTTTTTTTTTGTGAGTCTAA[G/C]TATTTATGTCCCTGTTAACTTATAA
- Phenotype
-
- Polymorphism
- G/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
BAGE2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79669132] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BAGE2
- Gene Name
- B melanoma antigen family member 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_182482.2 |
|
Intron |
|
|
NP_872288.2 |
- Gene
- LOC105378260
- Gene Name
- histone-lysine N-methyltransferase 2C
There are no transcripts associated with this gene.
View Full Product Details