Product Details

SNP ID
rs209023
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:10449827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGGTGGAGATGCCATACACTTT[T/C]GAACAACCAGATCTTGTGAGAACTC
Phenotype
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
BAGE2 PubMed Links

Gene Details

Gene
BAGE2
Gene Name
B melanoma antigen family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182482.2 Intron NP_872288.2

View Full Product Details