Product Details

SNP ID
rs28415497
Assay Type
Functionally Tested
NCBI dbSNP Submissions
1
Location
Chr.3:196581214 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCAGCTTTAGAATTTCCTTTTTT[C/T]TCTTTTTCCTTTTTTTTTTTTTTTT
Phenotype
MIM: 609112
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FBXO45 PubMed Links

Gene Details

Gene
FBXO45
Gene Name
F-box protein 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105573.1 Intron NP_001099043.1

View Full Product Details