Product Details
- SNP ID
-
rs2363991
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:48847358 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACTCTGGGGACTGTTGTGGGGTGG[A/G]GGGAGGGGGGAGAGAGAGCACTGGG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CDC14BL
PubMed Links
Gene Details
- Gene
- CDC14BL
- Gene Name
- CDC14 cell division cycle 14 C-like
There are no transcripts associated with this gene.
View Full Product Details