Product Details

SNP ID
rs17881537
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:104258600 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTACATAGACACTTCTCAGGCTG[C/G]GCATGGTAGCTCACGCCTATAACCC
Phenotype
MIM: 605482 MIM: 612314
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GSTO1 PubMed Links

Gene Details

Gene
GSTO1
Gene Name
glutathione S-transferase omega 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001191002.1 Intron NP_001177931.1
NM_001191003.1 Intron NP_001177932.1
NM_004832.2 Intron NP_004823.1
Gene
GSTO2
Gene Name
glutathione S-transferase omega 2
There are no transcripts associated with this gene.

Gene
MIR4482
Gene Name
microRNA 4482
There are no transcripts associated with this gene.

View Full Product Details