Product Details

SNP ID
rs34452287
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:87865477 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATTTAAGTATTGTTGTGATGCC[A/G]GAATTGCTGCTCACCGTTTTTAGGT
Phenotype
MIM: 612105 MIM: 601728
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLLN PubMed Links

Gene Details

Gene
KLLN
Gene Name
killin, p53-regulated DNA replication inhibitor
There are no transcripts associated with this gene.

Gene
PTEN
Gene Name
phosphatase and tensin homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000314.6 Intron NP_000305.3
NM_001304717.2 Intron NP_001291646.2
NM_001304718.1 Intron NP_001291647.1

View Full Product Details