Product Details

SNP ID
rs28689656
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:46549296 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCCAGGGGGCACGGAGCCCCCAC[T/C]GTCCCTGGCCCAGGTCTAGGACTAA
Phenotype
MIM: 611240
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
GPRIN2 PubMed Links

Gene Details

Gene
GPRIN2
Gene Name
G protein regulated inducer of neurite outgrowth 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014696.3 1576 UTR 3 NP_055511.2
XM_005277666.3 1576 UTR 3 XP_005277723.1
XM_011540388.2 1576 UTR 3 XP_011538690.1
XM_011540395.2 1576 UTR 3 XP_011538697.1
XM_011540400.2 1576 UTR 3 XP_011538702.1
XM_017016974.1 1576 UTR 3 XP_016872463.1
XM_017016975.1 1576 UTR 3 XP_016872464.1
XM_017016976.1 1576 UTR 3 XP_016872465.1
XM_017016977.1 1576 UTR 3 XP_016872466.1
XM_017016978.1 1576 UTR 3 XP_016872467.1
XM_017016979.1 1576 UTR 3 XP_016872468.1
XM_017016980.1 1576 UTR 3 XP_016872469.1
XM_017016981.1 1576 UTR 3 XP_016872470.1
XM_017016982.1 1576 UTR 3 XP_016872471.1
XM_017016983.1 1576 UTR 3 XP_016872472.1
XM_017016984.1 1576 UTR 3 XP_016872473.1
Gene
LOC107984026
Gene Name
arf-GAP with GTPase, ANK repeat and PH domain-containing protein 4-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017017029.1 1576 Intron XP_016872518.1
XM_017017030.1 1576 Intron XP_016872519.1
XM_017017031.1 1576 Intron XP_016872520.1
XM_017017032.1 1576 Intron XP_016872521.1
XM_017017033.1 1576 Intron XP_016872522.1
XM_017017034.1 1576 Intron XP_016872523.1
XM_017017035.1 1576 Intron XP_016872524.1
XM_017017036.1 1576 Intron XP_016872525.1
XM_017017037.1 1576 Intron XP_016872526.1
XM_017017038.1 1576 Intron XP_016872527.1
XM_017017039.1 1576 Intron XP_016872528.1
XM_017017040.1 1576 Intron XP_016872529.1

View Full Product Details