Product Details

SNP ID
rs35705883
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:52603929 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCTCCTGTTCTCTCTAGGGCAG[C/G]TCACCAGAGAGCCCCAGGTTTAGCG
Phenotype
MIM: 615784
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GPX7 PubMed Links

Gene Details

Gene
GPX7
Gene Name
glutathione peroxidase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015696.4 Intron NP_056511.2

View Full Product Details